Anti-MIA2 Rabbit Polyclonal Antibody

Supplier: Prosci
38-181
10098-344EA 576.13 USD
10098-344
Anti-MIA2 Rabbit Polyclonal Antibody
Antibodies
ELISA:<br>Indirect:<br>To detect hMIA-2 by indirect ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hMIA-2.<br><br>Sandwich<br>To detect hMIA-2 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our biotinylated Anti-Human MIA-2 as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hMIA-2. <br><br>Western Blot:<br>To detect hMIA-2 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hMIA-2 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: MIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR