Anti-WNT1 Rabbit Polyclonal Antibody

Supplier: Prosci
38-175
10098-332EA 576.13 USD
10098-332
Anti-WNT1 Rabbit Polyclonal Antibody
Antibodies
Wnt1 is a secreted protein that signals through the Frizzled family of cell surface receptors and is required for normal embryonic development. Wnt1 activation induces a complex signaling cascade that ultimately leads to the increased expression of over fifty genes. An important component of Wnt1 signaling is the stabilization, and resulting accumulation, of the intracellular signaling protein, beta catenine. Mature human Wnt1 is a glycosylated protein containing 343 amino acid residues.

ELISA:<br>Indirect:<br>To detect hWNT-1 by indirect ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hWNT-1.<br><br>Sandwich<br>To detect hWNT-1 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our biotinylated Anti-Human WNT-1 as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hWNT-1. <br><br>Western Blot:<br>To detect hWNT-1 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hWNT-1 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: WNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR