Anti-TNFSF15 Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
38-163
10098-310EA 576.13 USD
10098-310
Anti-TNFSF15 Rabbit Polyclonal Antibody (Biotin)
Antibodies
TL1A is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. It is a TNF like ligand for DR3 and TR6/DcR3 and functions as a T cell costimulator. TL1A is abundantly expressed in endothelial cells, but is not expressed in either B or T cells. The expression of this protein is inducible by TNF and IL 1 alpha. TL1A can activate NF kappa B and MAP kinases, and acts as an autocrine factor to induce apoptosis in endothelial cells and may function as an angiogenesis inhibitor. TL1A is a longer variant of TL1.

ELISA:<br>Direct:<br>To detect hTL-1A by direct ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hTL-1A.<br><br>Sandwich<br>To detect hTL-1A by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our polyclonal Anti-Human TL-1A as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hTL-1A. <br><br>Western Blot:<br>To detect hTL-1A by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hTL-1A is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: TNFSF15
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR