Anti-IL22 Rabbit Polyclonal Antibody

Supplier: Prosci
38-149
10098-286EA 576.13 USD
10098-286
Anti-IL22 Rabbit Polyclonal Antibody
Antibodies
A novel cytokine was recently identified in human and mouse and designated IL-21, which has significant homology to IL-2, IL-4, and IL-15. The receptor for IL-21 (IL-21R, also termed NILR for novel Interleukin receptor) is a new member of the class I cytokine receptor family. IL-21R forms a complex with the common cytokine receptor g chain, gc, and mediates IL-21 signaling. IL-21 and its receptor activate JAK-STAT signaling pathway. IL-21 is expressed in activated T cells, and HL-60 and THP-1 cell lines. IL-21 plays a role in the proliferation and maturation of NK, B and T cell populations.

ELISA:<br>Direct:<br>To detect mIL-22 by indirect ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mIL-22.<br><br>Sandwich<br>To detect mIL-22 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our biotinylated Anti-Murine IL-22 as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mIL-22. <br><br>Western Blot:<br>To detect mIL-22 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mIL-22 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: Il22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR