Anti-RETNLA Rabbit Polyclonal Antibody

Supplier: Prosci
38-147
10098-282EA 576.13 USD
10098-282
Anti-RETNLA Rabbit Polyclonal Antibody
Antibodies
A family of resistin-like molecules (RELMs) has been identified in rodents and humans. RELM alpha belongs to a unique family of tissue-specific cytokines termed FIZZ (found in inflammatory zone) and RELM. The three known members of this family; Resistin, RELM alpha and RELM beta are 85-94 amino acid secreted proteins sharing a conserved C-terminal domain. RELM alpha and Resistin are secreted exclusively by adipocytes while RELM beta is expressed in the epithelium of the colon and small bowel. The RELMs together with resistin comprise a class of tissue-specific signaling molecules. The physiological role and molecular targets of RELM alpha are still unknown.

ELISA:<br>Indirect:<br>To detect mRELMa by indirect ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mRELMa.<br><br>Sandwich<br>To detect mRELMa by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our biotinylated Anti-Murine RELMa as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mRELMa. <br><br>Western Blot:<br>To detect mRELMa by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mRELMa is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: Retnla
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR