Anti-CCL24 Goat Polyclonal Antibody

Supplier: Prosci
38-116
10097-902EA 576.13 USD
10097-902
Anti-CCL24 Goat Polyclonal Antibody
Antibodies
ELISA:<br>Indirect:<br>To detect hEotaxin-2 by indirect ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hEotaxin-2.<br><br>Sandwich<br>To detect hEotaxin-2 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our biotinylated Anti-Human Eotaxin-2 as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hEotaxin-2. <br><br>Western Blot:<br>To detect hEotaxin-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hEotaxin-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: CCL24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR