Anti-CTGF Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
38-105
10098-202EA 576.13 USD
10098-202
Anti-CTGF Rabbit Polyclonal Antibody (Biotin)
Antibodies
CTGF (connective tissue growth factor) is a 38kDa, cysteine-rich, secreted peptide. It is a new member of the peptide family that include serum-induced immediate early gene products, a v-src-induced peptide and a putative proto-oncogene. Among the many functions of the CTGF gene family are embryogenesis, wound healing and regulation of extracellular matrix production.

ELISA: Direct: To detect hCTGF by direct ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hCTGF. Sandwich: To detect hCTGF by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. Western Blot: To detect hCTGF by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hCTGF is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

CTGF antibody was purified by affinity chromatography and then biotinylated.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR