Anti-NGFR Rat Monoclonal Antibody [clone: Nagy-1]

37-146
10070-774EA 550.52 USD
10070-774
Anti-NGFR Rat Monoclonal Antibody [clone: Nagy-1]
Flow Cytometry (1:1,000)

PBS containing 0.02% sodium azide

NGF antibody can be stored at 4°C, stable for one year.

Type: Primary
Antigen: NGFR
Clonality: Monoclonal
Clone: Nagy-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR