Anti-SARM1 Mouse Monoclonal Antibody [clone: Sarmy-1]

Supplier: Prosci
37-145
10070-772EA 740.9 USD
10070-772
Anti-SARM1 Mouse Monoclonal Antibody [clone: Sarmy-1]
Antibodies
Immunoprecipitation (1:200)

PBS containing 10% glycerol and 0.02% sodium azide

SARM antibody can be stored at 4°C, stable for one year.

Type: Primary
Antigen: SARM1
Clonality: Monoclonal
Clone: Sarmy-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR