Anti-CRADD Mouse Monoclonal Antibody [clone: Raiddy-1]

37-142
10070-766EA 728.22 USD
10070-766
Anti-CRADD Mouse Monoclonal Antibody [clone: Raiddy-1]
Antibodies
Immunoprecipitation (1:100) Western Blot (1:1,000)

PBS containing 0.02% sodium azide

RAIDD antibody can be stored at 4°C, stable for one year.

Type: Primary
Antigen: CRADD
Clonality: Monoclonal
Clone: Raiddy-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR