Anti-Staph enterotoxin Mouse Monoclonal Antibody [clone: 7935]

Supplier: Prosci
35-693
10069-402EA 665.97 USD
10069-402
Anti-Staph enterotoxin Mouse Monoclonal Antibody [clone: 7935]
Antibodies
Staph enterotoxin antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Staph enterotoxin antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Staph enterotoxin Antibody [7935]
Clonality: Monoclonal
Clone: 7935
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR