Anti-PAGA Mouse Monoclonal Antibody [clone: 7821]

Supplier: Prosci
35-685
10069-392EA 665.97 USD
10069-392
Anti-PAGA Mouse Monoclonal Antibody [clone: 7821]
Antibodies
Anthrax (B. anthracis) antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Anthrax antibody (B. anthracis) is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Anthrax (B. anthracis) Antibody [7821]
Clonality: Monoclonal
Clone: 7821
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR