Anti-Diphtheria toxin Mouse Monoclonal Antibody [clone: 7801]

Supplier: Prosci
35-683
10069-388EA 665.97 USD
10069-388
Anti-Diphtheria toxin Mouse Monoclonal Antibody [clone: 7801]
Antibodies
Diphtheria toxin antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Diphtheria toxin antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Diphtheria toxin Antibody [7801]
Clonality: Monoclonal
Clone: 7801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR