Anti-Canine Distemper Virus Mouse Monoclonal Antibody [clone: 7515]

Supplier: Prosci
35-670
10069-374EA 665.97 USD
10069-374
Anti-Canine Distemper Virus Mouse Monoclonal Antibody [clone: 7515]
Antibodies
Canine Distemper Virus antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Canine Distemper Virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Canine Distemper Virus Antibody [7515]
Clonality: Monoclonal
Clone: 7515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR