Anti-Feline Leukemia virus Mouse Monoclonal Antibody

35-664
10069-362EA 381.42 USD
10069-362
Anti-Feline Leukemia virus Mouse Monoclonal Antibody
Feline Leukemia virus antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Feline Leukemia virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Feline Leukemia virus
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR