Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody

35-622
10070-550EA 381.42 USD
10070-550
Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody
Neisseria gonorrhoeae antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Neisseria gonorrhoeae antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Neisseria gonorrhoeae
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR