Anti-Rabies Virus Mouse Monoclonal Antibody [clone: 6041]

Supplier: Prosci
35-618
10070-542EA 665.97 USD
10070-542
Anti-Rabies Virus Mouse Monoclonal Antibody [clone: 6041]
Antibodies
Rabies Virus antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Rabies Virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Rabies Virus Antibody [6041]
Clonality: Monoclonal
Clone: 6041
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR