Anti-Aspergillus Antibody [clone: 5141]

35-595
10070-526EA 633.26 USD
10070-526
Anti-Aspergillus Antibody [clone: 5141]
Antibodies
Aspergillus antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Aspergillus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Aspergillus Antibody [5141]
Clonality: Monoclonal
Clone: 5141
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgM
Reactivity: Fungus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR