Anti-HSV Mouse Monoclonal Antibody [clone: 2002]

Supplier: Prosci
35-552
10069-282EA 665.97 USD
10069-282
Anti-HSV Mouse Monoclonal Antibody [clone: 2002]
Antibodies
Herpes Virus (type 6) antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Herpes Virus antibody (type 6) is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Herpes Virus (type 6) Antibody [2002]
Clonality: Monoclonal
Clone: 2002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR