Anti-HIV Mouse Monoclonal Antibody [clone: 1981]

Supplier: Prosci
35-549
10071-568EA 665.97 USD
10071-568
Anti-HIV Mouse Monoclonal Antibody [clone: 1981]
Antibodies
HIV antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

HIV antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HIV Antibody [1981]
Clonality: Monoclonal
Clone: 1981
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR