Anti-Influenza Mouse Monoclonal Antibody [clone: 1131]

Supplier: Prosci
35-462
10069-166EA 665.97 USD
10069-166
Anti-Influenza Mouse Monoclonal Antibody [clone: 1131]
Antibodies
Influenza B virus antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Influenza B virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Influenza B virus Antibody [1131]
Clonality: Monoclonal
Clone: 1131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR