Anti-Streptococcus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: ProSci
35-194
10070-778EA 701.05 USD
10070-778
Anti-Streptococcus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Purified IgG fraction of antiserum covalently coupled with high purity Isomer I of fluorescein isothiocyanate. Care is taken to insure complete removal of any free fluorescein from the final product.

Group A Streptococcus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Group A Streptococcus Antibody (FITC)
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR