Anti-Feline Leukemia virus Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: ProSci
35-187
10069-640EA 665.97 USD
10069-640
Anti-Feline Leukemia virus Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.

Feline Leukemia Virus antibody is in a phosphate saline buffer with BSA (10mg/ml) and preserved with 0.002% thimerosal.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Feline Leukemia Virus Antibody (HRP)
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR