Anti-Feline Leukemia virus Goat Polyclonal Antibody

Supplier: ProSci
35-185
10069-636EA 576.12 USD
10069-636
Anti-Feline Leukemia virus Goat Polyclonal Antibody
Antibodies
Purified IgG fraction of antiserum to a purity of >90%.

Feline Leukemia Virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Feline Leukemia Virus Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR