Anti-DNAAF1 Rabbit Polyclonal Antibody

Supplier: ProSci
30-866
10110-832EA 745.96 USD
10110-832
Anti-DNAAF1 Rabbit Polyclonal Antibody
Antibodies
LRRC50 contains 6 LRR (leucine-rich) repeats. It is proposed that LRRC50 to be a novel candidate gene for human cystic kidney disease, involved in regulation of microtubule-based cilia and actin-based brush border microvilli.

LRRC50 antibody can be used for detection of LRRC50 by ELISA at 1:1562500. LRRC50 antibody can be used for detection of LRRC50 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DNAAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR