Anti-EXOSC4 Rabbit Polyclonal Antibody

Supplier: Prosci
29-451
10108-254EA 680.93 USD
10108-254
Anti-EXOSC4 Rabbit Polyclonal Antibody
Antibodies
EXOSC4 belongs to the RNase PH family. It is a component of the exosome 3'->5' exoribonuclease complex and is required for the 3' processing of the 7S pre-RNA to the mature 5.8S rRNA. It has a 3'-5' exonuclease activity.

EXOSC4 antibody can be used for detection of EXOSC4 by ELISA at 1:62500. EXOSC4 antibody can be used for detection of EXOSC4 by western blot at 5.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EXOSC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR