Anti-ZNF341 Rabbit Polyclonal Antibody

Supplier: Prosci
27-266
10104-642EA 680.93 USD
10104-642
Anti-ZNF341 Rabbit Polyclonal Antibody
Antibodies
Zinc Finger Protein 341 is a new candidate transcription factor.

ZNF341 antibody can be used for detection of ZNF341 by ELISA at 1:62500. ZNF341 antibody can be used for detection of ZNF341 by western blot at 2.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZNF341
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR