Anti-TFAP2B Rabbit Polyclonal Antibody

Supplier: Prosci
27-112
10100-576EA 745.96 USD
10100-576
Anti-TFAP2B Rabbit Polyclonal Antibody
Antibodies
TFAP2B belongs to the AP-2 family which is developmentally regulated and have distinct overlapping functions in the regulation of many genes governing growth and differentiation. TFAP2B binds DNA as a dimmer and can form homodimers or heterodimers with other AP-2 family members. It may be a candidate for conferring susceptibility to type 2 didabetes. This gene encodes a member of the AP-2 family of transcription factors. AP-2 proteins form homo- or hetero-dimers with other AP-2 family members and bind specific DNA sequences. They are thought to stimulate cell proliferation and suppress terminal differentiation of specific cell types during embryonic development. Specific AP-2 family members differ in their expression patterns and binding affinity for different promoters. This protein functions as both a transcriptional activator and repressor. Mutations in this gene result in autosomal dominant Char syndrome, suggesting that this gene functions in the differentiation of neural crest cell derivatives. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-144 AU141084.1 1-144 145-1684 BC037225.1 1-1540 1685-5370 AL049693.16 11928-15613 5371-5770 BU738725.1 18-417 c

  • TFAP2B antibody can be used for detection of TFAP2B by ELISA at 1:62500. TFAP2B antibody can be used for detection of TFAP2B by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TFAP2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR