Anti-ZNF407 Rabbit Polyclonal Antibody

Supplier: Prosci
25-119
10101-550EA 745.96 USD
10101-550
Anti-ZNF407 Rabbit Polyclonal Antibody
Antibodies
ZNF407 contains 22 C2H2-type zinc fingers. ZNF407 may be involved in transcriptional regulation.

ZNF407 antibody can be used for detection of ZNF407 by ELISA at 1:312500. ZNF407 antibody can be used for detection of ZNF407 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZNF407
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR