Anti-ZC2HC1A Rabbit Polyclonal Antibody

Supplier: Prosci
25-043
10101-394EA 745.77 USD
10101-394
Anti-ZC2HC1A Rabbit Polyclonal Antibody
Antibodies
The exact function of FAM164A (C8orf70) remains unknown.

C8orf70 antibody can be used for detection of C8orf70 by ELISA at 1:62500. C8orf70 antibody can be used for detection of C8orf70 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZC2HC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR