Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BOD Measurement, mercury-free, OxiTop® Control 6/12

Supplier: WTW MEASUREMENT SYSTEMS INC
OxiTop®
208201 208204
  • Standard and routine measurements of 6 or 12 samples in parallel with automatic data transfer
  • Controller has a range of between 0...40/80/200/400/800/2000/4000 mg/l BOD
  • Automatic assignment of a sample identification number
  • Measurement period: 0,5 h to 99 days
  • Heads can store up to 360 data sets

The automated system for measuring BOD, OxiTop® Control, consists of an OxiTop® Control measuring head and an infrared controller. This system is exceptionally easy to use. The controller is capable of configuring up to 100 measuring heads and receiving the measurement data via infrared transmission. Each sample is easily identified and recognised by the controller thanks to the measuring head, each of which is associated with a unique serial number. The controller is multilingual to make it as easy as possible to operate. The digital display is clear and offers step by step guidance for all user operations.

Complete package for 6 simultaneous measurements with  OxiTop® OC 100 Controller,and IS 6 Inductive Stirring System for mains operation 230 V/50/60 Hz and 6 OxiTop® C measuring heads and appropriate standard accessories.

Complete package for 12 simultaneous measurements with  OxiTop® OC 100 Controller,and IS 12 Inductive Stirring System for mains operation 230 V/50/60 Hz and 12 OxiTop® C measuring heads and appropriate standard accessories.




 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
OxiTop® Control 6 208201 80085-450 Each Retrieving Restricted
OxiTop® Control 12 208204 80085-452 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
OxiTop® Controller OC110 Set for the control of OxiTop® Control measuring systems with PC evaluation and documentation. Includes PC communication software Achat OC and interface cable AK 540/B 208209 80086-720 Each Retrieving Restricted
OxiTop® Controller OC110 for the control of BOD and special applications of OxiTop® Control measuring systems G208207 80085-454 Each Retrieving Restricted
OxiTop® Controller OC100 for the control of routine and standard OxiTop®-C BOD measuring systems 208208 97042-944 Each Retrieving Restricted
OxiTop® head, replacement (yellow ) 208810 80085-472 Each Retrieving Restricted
OxiTop®-6 heads, replacement (3x yellow + 3x green) 208812 80085-474 Each Retrieving Restricted
Bottle MF 45/1000 ml, 2 side connectors 209108 80085-486 Each Retrieving Restricted
Bottle MF 45/250 ml, 2 side connectors 209106 80085-484 Each Retrieving Restricted
Measuring vessel 1000 ml for OxiTop®-C with accessories 1×DV/MG adapter with rack and gasket, 2 x rubber sleeves GK 600, 4×DV/MG/KL clamps and 1×MB50 measuring beaker (50 ml) 209019 80085-482 Each Retrieving Restricted
OxiTop® BOD Respirometer Systems thermostat box, for up to 12 OxiTop® units and stirrer base, 115 V, 50 Hz 208433 80085-470 Each Retrieving Restricted
OxiTop® Control 2 heads, replacements 208823 80085-478 Each Retrieving Restricted
OxiTop® Control 6 heads, replacements 208822 80085-476 Each Retrieving Restricted
OxiTop® Control head, replacement 208830 80085-480 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty