Allyl cyanide ≥92.0%

Supplier: TCI America

A0227-25ML
TCA0227-25MLEA 42.67 USD
TCA0227-25ML
Allyl cyanide ≥92.0%
Allyl cyanide

CAS Number: 109-75-1
MDL Number: MFCD00001962
Molecular Formula: C4H5N
Molecular Weight: 67.09
Purity/Analysis Method: 92.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 24
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3-Butenenitrile

Formula: C₄H₅N
Boiling Pt: 118 °C
Density: 0.83 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00001962
CAS Number: 109-75-1
UN: 1992
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/76/57/28927657.jpg

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR