2,2,4-Trimethyl-1,3-pentanediol diisobutyrate ≥98.5%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,2,4-Trimethylpentane-1,3-diyl bis(2-methylpropanoate)

T1366-1LT T1366-20LT T1366-4LT
700004-954EA 197.42 USD
700004-954 700004-952 700004-950
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate ≥98.5%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate

2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate is used as a plastizer and viscosity control agent in cosmetic and personal care products.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₃₀O₄
MW: 286.41 g/mol
Density: 0.941
Storage Temperature: Ambient
MDL Number: MFCD00059267
CAS Number: 6846-50-0

Order Now

Specification Test Results

Molecular Weight 286.41

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR