Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BRAND® Ultra Low Retention 50 µL Pipette Tips, BrandTech®

Supplier: BRANDTECH SCIENTIFIC INC.
732326 732306
BRAND Ultra-Low Retention pipette tips are produced through a patented physicochemical prodess that provides a homogeneous defect-free surface

BRAND 50 microliter pipette tips are a longer style (50mm) ultra-micro/crystal tip for use on many pipettes with a nominal/maximum volume less than 20 microliters. Extended volume allows reverse pipetting of viscous or volatile liquids for greater accuracy. Graduations at 2.5, 10, 25 and 50 microliters allow easy referencing of pipetted volume.

All tips are produced under cleanroom conditions. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes, and have 20% less waste. Sterile TipRefills have an included transfer aid to ensure sterility.

This results in an extremely low surface tesnion, over 50% less than PTFE. This significantly reuces sample loss and improves reproducibility. BRAND Ultra Low Retention tips have no additives than can leach out, or coatings to wash off. Fully autoclavable at 121 C without damage to surface properties.

Certifications: CE-IVD

Ordering information: All rack-packed tips are DNA-, RNase-, ATP-, and endotoxin-free.



 

ORDER

Expand All / Collapse All
Volume Sterility Packed Supplier No. VWR Catalog Number Unit Price Quantity
1–50 µL Nonsterile 5 TipBoxes 732306 10034-088 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
1–50 µL Sterile 10 TipBoxes 732326 10034-046 Pack of 960 Retrieving
Product and/or packaging is recyclable or reduces waste
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging is recyclable or reduces waste

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty