BRAND filtered pipette tips for pipettes using 200 ul tips are an excellent choice to reduced contaminating aerosols when using pipettes with a yellow color code.
- Non-sealing filters made of polyetheylene
- Sample recovery possible if filter is wetted
- Autoclavable at 121˚C for 20 minutes
- Environmentally-friendly packaging
All tips are produced under cleanroom conditions. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes, and have 20% less waste. Sterile TipRefills have an included transfer aid to ensure sterility.
Three different designs are available: a 2 to 20 µl size; a 5 to 100 µl size, and a 5 to 200 µl size. Non-self-sealing filters allow easy recovery of sample if filter is wetted. PE filters are free from additives.
Certifications: CE-IVD.
Ordering information: All rack-packed tips are DNA-, RNase-, ATP-, and endotoxin-free.