Beta Block with Base and Cover, Acrylic, Dynalon
172334
71070-144EA 135.7 USD
71070-144
Beta Block with Base and Cover, Acrylic, Dynalon
Heavy solid acrylic blocks hold four 1.5ml microcentrifuge tubes
Designed for both bench-top work and long term storage of beta-emitting radioactive isotope samples. Supplied with cover, non-skid feet and radiation hazard warning labels.
Ordering information: Additional sizes and materials are available upon request.