Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Versilic® SPX-50 High Strength Silicone Tubing, Saint Gobain

Supplier: SAINT GOBAIN PERFORMANCE PLST
ABX00017 ABX00023
  • Provides resiliency
  • Long life and durability
  • Ultra-smooth inner bore reduces risk of particle entrapment
  • Taste and odor free
  • Withstands temperature extremes from -75oF to 350oF

Peroxide-cured Versilic® Silicone Tubing Formulation SPX-50 is designed for use in applications where flexibility, resiliency, and durability are required. Produced from a proprietary combination of silicone elastomers, Versilic® Silicone Tubing optimizes critical physical properties such as tensile strength, elongation, and compression set, resulting in a more physically durable product. Its smooth inner surface reduces the risk of particulate entrapment and microscopic buildup during fluid transfer. Every lot of raw material used in the manufacture of Versilic® Silicone Tubing undergoes a series of in-house physical property testing before use. This stringent evaluation of ingredients helps to provide the repeatable quality found in every foot of Versilic® Silicone Tubing. Annual testing on actual tubing samples is performed for reaffirmation of adherence to USP Class VI criteria. Heavy metals extraction analysis, as well as, cytotoxicity testing is performed to confirm the tubing's biocompatibility. Versilic® Silicone Tubing complies fully with the requirements of the USP Class VI Criteria and is entirely non-toxic, non-hemolytic and non-pyrogenic. Versilsic® Silicone Tubing provides versatility in use for a broad range of applications.

Meets USP Class VI and FDA 21 CFR Part 177.2600 criteria, 3-A Sanitary Standards, NSF 51 Standard

Ordering information: Length of package is 50 feet of translucent tubing.



 

ORDER

Expand All / Collapse All
I.D. x O.D. Wall Thickness Working Pressure Bend Radius Supplier No. VWR Catalog Number Unit Price Quantity
1/4 x 3/8 1/16" 9.0 psi 3/4" ABX00017 89030-120 Pack of 2 Retrieving
5/16 x 1/2 3/32" 10.0 psi 5/8" ABX00023 89030-122 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Durometer Hardness 50 (Shore A)

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty