Would you like to log in again?
UVP Economy Photo Documentation System, Analytik Jena
Supplier: Analytik Jena US
97-0274-04
Allows the researcher to view gels with UV transilluminator in darkened environment provided by a mini hood. A variety of configurations are available as listed below. The DS-34 fixed, focal length camera, along with a transilluminator and darkroom hood, provide the convenience and advantages of documentation at the lab benchtop. Variable shutter speeds and aperture settings allow the user to set the proper exposure for specific documentation. System includes a DS-34 camera, UV transilluminator, darkroom hood, filter 22, UV blocking spectacles, Gel-cutter tool, and instant B&W film.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
PhotoDoc-It w/M-20 Transilluminator 97-0274-04 95026-906 Each Retrieving
Product available on GSA Advantage® Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty