Boekel Microtube ThermoShaker
270590 270591
97004-382EA 187.67 USD
97004-382 97004-384
Boekel Microtube ThermoShaker
This benchtop shaker/incubator provides excellent uniformity with a variable speed of 250 to 1400 rpms. With interchangeable platforms, this unit accommodates your needs in the life science lab. Interchangeable platforms to meet your tube needs.