SP Bel-Art Write-On™ Label Tape Dispensers, Bel-Art Products, a part of SP
H13461-0000
490014-302EA 107.89 USD
490014-302
SP Bel-Art Write-On™ Label Tape Dispensers, Bel-Art Products, a part of SP
Use one hand and the Write-On labeling tape dispenser to quickly make a label for your sample without fumbling.
  • Label tape dispensers are compatible with rolls of various widths and color
  • Practical writing platform accommodates left- and right-handed users
  • Lighter model can be attached to vertical surfaces
  • Deluxe model features a non-skidding weighted steel base
Dispensers hold up to 7 rolls of labeling tape of various colors and widths to meet your individual labeling needs
The epoxy-coated steel platform has finger holes for releasing the tape with a cut off edge for one-hand use. A weighted base with non-skid pads prevents sliding or lifting (4" x 6" platform). Deluxe Tape Dispenser has a plastic-coated, weighted steel base with rubber feet to prevent slipping on the benchtop when pulling the labeling tape. The serrated cutting edge is steel for long term use. 12" x 5-5/8" x 4-3/4" (H)
To get started, dispensers are supplied with a roll of white (L-2001-1) Write-On Tape. Economy Tape Dispenser is designed for lighter duty use as the plastic cutting edge is molded into the housing. There is a series of mounting holes in the unit for secure attachment with screws (not supplied) to the benchtop. For a space-saving alternative, this dispenser can also be attached to many vertical surfaces. Single-Roll Tape Dispenser has a practical writing platform designed to accommodate both left- and right-handed users for labeling tape prior to dispensing.