Would you like to log in again?
Sorenson Gold Series Semi-skirted 96-well PCR Plates: FastPlate for Real Time PCR
Supplier: SORENSON BIOSCIENCE
3880-E
  • Optimized for the new Applied Biosystems' Fast PCR Systems
  • PCR results in 25 minutes when used with the 9800, Real-Time PCR results in 40 minutes when used with the 7500
  • Low profile and thin-wall design reduce cycle times
  • Ultra-clear wells produce optimal signal intensity
  • Fits most popular thermal cyclers
  • Working volume = 200 L
Ideal for running less than 96 samples on your cycler. These PCR plates have a super thin wall for rapid and efficient thermal transfer. Uniform well thickness ensures well-to-well and plate-to-plate uniformity. Molded from ultra clear polypropylene, these plates are available in an array of colors. All plates are certified DNase-, RNase-, and pyrogen-free. The new 96-well FastPlate provides optimal performance on the new Fast PCR systems available from Applied Biosystems. The low profile design and thin walls reduce cycle times to as short as 25 minutes for a standard PCR reaction. The ultra-clear wells produce optimal signal intensity and feature 200 L working volume. The FastPlate is also compatible with most popular 96-well thermal cyclers.
Thermal cycler/PCR system compatibility:
Applied Biosystems®
9800 Fast PCR Thermal Cycler - Recommended
7500 Real-time PCR System - Recommended
7900HT Fast Real-time PCR System - Recommended
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
FastPlate 96-well, Natural 3880-E 490014-860 Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty