Would you like to log in again?
Non-skirted, Chimney-well PCR Plates
Supplier: BioExpress
1721
  • Alphanumeric-printed PCR plates
  • Thin wall design to ensure uniform, rapid thermal transfer
  • 100% quality control on each well of every plate
  • Certified RNase/DNase-free, non-pyrogenic, and human DNA/PCR inhibitors-free
MegaBACE 1000 certified. The MBplate features a raised rim on each tube for eliminating well-to-well contamination. Molecular Dynamics certifies the MBplate for use on the MegaBACE 1000 DNA Analysis System. Its standard 96-well footprint lends it to complete automated sample handling. A thin-wall design reduces cycle times and a grid reference simplifies sample identifications and plate orientation. MegaBACE is a trademark of Molecular Dynamics, Inc. PCR Process is covered by patents owned by Hoffman-LaRoche, Inc. MULTIMaxLiquid Handling Products.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Color Supplier No. VWR Catalog Number Unit Price Quantity
8-Strip Caps for MBplate Natural 1721 490015-006 Each Retrieving Blocked
Pack of 1,000 Retrieving
Blocked This product has been blocked by your organization.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty