Would you like to log in again?
Extended Reach 1000 µl Blue Tips
Supplier: SCIENTIFIC SPECIALTIES, INC.
4331-01 4330-01-120IS
This blue tip is a highly polished tip which offers several advantages over a standard tip
As seen in the illustration, the tip is narrower to enable you to reach the bottom of many tubes, including a 12 x 75 mm culture tube. Also, the tip is longera full 3 inches long. This tip has the same excellent fit as all of our GeneMate tips.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
200-1000 µl, Bulk 4330-01-120IS 490015-164 Bag of 1,000 Retrieving
Case of 10000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty