Would you like to log in again?
Large Orifice Tips
Supplier: SCIENTIFIC SPECIALTIES, INC.
For viscous samples, proteins, genomic DNA, and other delicate samples, GeneMate Large Orifice Tips are designed specifically for work with viscous samples that can get held up in a standard 200 L tip. Genomic DNA, hybridomas, proteins, and cell suspensions can be damaged by shearing forces associated with pipetting. Our Large Orifice tips reduce these shearing effects substantially.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Wide Orifice 250 µl Tips 4290-00-120IS 490016-294 Bag of 1,000 Retrieving
Case of 10000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty