Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MBP BreakThru Pipet Tips

Supplier: Thermo Scientific
3552-50 3551-50 2149P-50
  • Universal to all 8 x 12 pipet tip trays
  • Works with ART self sealing barrier pipet tips
  • No snaps, buttons or machines needed
  • Uses 85% less packaging and 53% less space
  • 96 tips per insert / 10 inserts per pack / 10 racks per case
  • Recyclable Packing
  • BreakThru Packaging

BreakThru Technology. This revolutionary packaging design allows us to bring you an environmentally friendly alternative to the packaging of ART® (Aerosol Resistant Tips) and MBP® brand pipet tips. With BreakThru Technology, MBP continues to deliver peace of mind by providing the highest quality tips with the least amount of environmental impact. This new environmental packaging system uses no insert because of our unique BreakThru transfer plate. This exciting new technology allows you to use any existing 8 x 12 tip trays in your lab, making it not only the first refill system universal to all manufacturers’ pipet tip trays, but the first to work with ART self-sealing barrier tips.

BreakThru packaging means less waste and more room! BreakThru packaging is 100% recyclable and can be disposed of with your standard recyclable waste. It uses 53% less bench space and 85% less packaging material, increasing your workspace and reducing the environmental impact of your research. BreakThru packaging means more efficiency. There are no mechanical moving parts so it works consistently and is very reliable. There are no snaps, buttons, or springs required to load the tips so it loads a full 96 tips into the rack faster and with fewer steps.




 

ORDER

Expand All / Collapse All
Description Volume Sterility Supplier No. VWR Catalog Number Unit Price Quantity
Universal Micro Point Tip, Racked 200 µL Nonsterile 3552-50 490015-850 Box of 960 Retrieving
Case of 9600 Retrieving
Universal Micro Point Tip, Racked, Certified Pure 200 µL Sterile 3551-50 490016-338 Box of 960 Retrieving
Case of 9600 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty