SpeedLoader™ System
Supplier: Medax International
03R 3211C 5211C
490015-134BX 55.89 USD
490015-134 490016-334 490014-534
SpeedLoader™ System
Pipette Tips
These tips are manufactured from 100% low retention polypropylene and are universal fit, metal free, and feature a beveled tip.
  • Pipette tip and racking system
  • Ultra-low fluid retention
  • Thin wall for high accuracy
  • Universal fit
  • Designed for research
  • Soft collar for secure seal
This Speed loader system is designed to work with your existing 96-place pipette tip racks. This system saves 75% on plastic used by conventional tip and rack systems. Storage is easy and the SpeedLoader System saves 70% on tip storage space. Starter units contain 10 racks of tips and one SpeedLoader Loading System.
Delivery information: Each case includes a transfer device which allows you to use your existing racks and load 96 tips within seconds.