Would you like to log in again?
Multichannel Disposable Solution Basin
Supplier: Heathrow Scientific
HS20521C HS20521B HS20521A
Constructed of white polystyrene or a clear PVC, the disposable basins have angled sides and V-shaped bottoms allowing nearly complete sample withdrawal.
ORDER
Expand All / Collapse All
Description Material Color Supplier No. VWR Catalog Number Unit Price Quantity
55 mL Multichannel Solution Basin PS White HS20521B 490015-114 Box of 100 Retrieving
55 mL Multichannel Solution Basin PVC Clear HS20521A 490015-714 Box of 100 Retrieving
55 mL Multichannel Solution Basin, Sterile PVC White HS20521C 490015-116 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty