Robotic Tips for FX Robotic Workstations
4612-SF-110IS 4622-00-110IS 4612-S0-110IS 4632-S0 4622-SF-110IS 4632-00-110IS 4612-00-110IS
490015-172BX 90.92 USD
490015-172 490015-692 490016-412 490015-694 490014-536 490015-844 490014-538
Robotic Tips for FX Robotic Workstations
Robotic pipette tips are available in racked, filter, and sterile versions for the Beckman 1000 and 2000 robotic workstations.
  • 1-250 uL FX/Multimek 96/Cyberlab-type Tip Racked, Natural
  • Polypropylene
  • Case Dimensions (approx) 699 x 451 x 289 mm
  • Case Weight (approx) 20 kg
  • Lot-by-lot Certification
  • Proven on OEM Equipment
  • Contamination Free
Tips are in special racks with lift-off cover. They have separate compartments for each tip to eliminate potential for cross contamination.