Would you like to log in again?
Transfer Pipets
Supplier: Globe Scientific
  • Ideal for liquid handling & transfer applications
  • Non-toxic and chemically inert
  • Can be heat sealed to use for liquid storage
  • Can be frozen in liquid nitrogen
  • Low-density polyethylene (LDPE)
  • Can be gas (EtO) sterilized
Ordering information: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean. Each dispenser box contains 400 pipets.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Capacity Length Bulb Draw Stem Diameter Tip Size Drops per mL Temperature Range Supplier No. VWR Catalog Number Unit Price Quantity
Standard Pipet 7 mL 155 mm (6.1") 3.2 mL 7.8 mm (0.31") 3.1 mm (0.12") 20 -196°C to 90°C 138080 490016-614 Case of 5000 Retrieving
Pack of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty