Sterile Saline Solution (NaCl), 0.9%
MSDW-1000GM
490015-454EA 25.51 USD
490015-454
Sterile Saline Solution (NaCl), 0.9%
Comes in 1000 mL autoclavable polycarbonate bottle
Recipe: 9 g/L. Store at room temperature. Shelf life is three years from date of manufacture.